Stampa…
4'-Methoxy-2-chloroacetophenone
Fornitore: TCI EUROPE
Synonyms: 2-Chloro-1-(4-methoxyphenyl)ethanone, 4-Methoxyphenacyl chloride
M3268-25G M3268-5G
TCIAM3268-25GEA 369 EUR
TCIAM3268-25G TCIAM3268-5G
4'-Methoxy-2-chloroacetophenone
4'-Methoxy-2-chloroacetophenone
Formula: C₉H₉ClO₂
Peso molecolare: 184,6196 g/mol
MDL Number: MFCD00216508
CAS Number: 2196-99-8
UN: 3261