Stampa…
4,5,6,7-Tetrachloro-2-hydroxyisoindoline-1,3-dione
Fornitore: TCI EUROPE
Synonyms: N-Hydroxytetrachlorophthalimide
H1765-1G H1765-5G
TCIAH1765-1GEA 27 EUR
TCIAH1765-1G TCIAH1765-5G
4,5,6,7-Tetrachloro-2-hydroxyisoindoline-1,3-dione
4,5,6,7-Tetrachloro-2-hydroxyisoindoline-1,3-dione
Formula: C₈HCl₄NO₃
Peso molecolare: 300,91 g/mol
MDL Number: MFCD29088000
CAS Number: 85342-65-0