Stampa…

4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine

Fornitore: TCI

B6237-1G B6237-5G
TCIAB6237-1GEA 56 EUR
TCIAB6237-1G TCIAB6237-5G
4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine
4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine
Formula: C₆H₅BrN₄
MDL Number: MFCD09033848
CAS Number: 937046-98-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR