Stampa…
Azido-dT phosphoramidite
Fornitore: AAT BIOQUEST INC
6217 6216
AATB6217EA 321 EUR
AATB6217 AATB6216
Azido-dT phosphoramidite
Azido-dT phosphoramidite
Azido-dT Phosphoramidite is an excellent building block for introducing an azide group into an oligo via dT.
  • Stable for 6 months
Quality tested in-house