Stampa…
Diisobutylaluminium hydride 1.0 M in heptane, AcroSeal®
Fornitore: Thermo Scientific
Pericolo
Synonyms: Di-i-butylaluminum hydride, DIBAL-H
201051000 201058000
201051000.EA 80.5 EUR
201051000. 201058000.
Diisobutylaluminium hydride 1.0 M in heptane, AcroSeal®
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
Peso molecolare: 142,22 g/mol
Flash Pt: –4 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3399
ADR: 4.3,I
Visualizza i codici
Appearance Clear solution
Strength 19.5 to 20.5 % (DIBAL-H, on Al basis)