Stampa…

Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (PE (Phycoerythrin))

Fornitore: AAT BIOQUEST INC
100521L0 100521L1 100521L2
AATB100521L0EA 271 EUR
AATB100521L0 AATB100521L1 AATB100521L2
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (PE (Phycoerythrin))
Anticorpi
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Attenzione: For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR