Stampa…

Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 710)

Fornitore: AAT BIOQUEST
100520K0 100520K1
AATB100520K0EA 371 EUR
AATB100520K0 AATB100520K1
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 710)
Anticorpi
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Attenzione: For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: iFluor® 710
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR