Stampa…
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 860)
Fornitore: AAT BIOQUEST
100320R0 100320R1
AATB100320R0EA 371 EUR
AATB100320R0 AATB100320R1
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 860)
Anticorpi
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kDa member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Attenzione: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: iFluor® 860
Epitope:
Host: Mouse
Isotype:
Reactivity: Human