Stampa…

Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 810)

Fornitore: AAT BIOQUEST
100210O0 100210O1
AATB100210O0EA 371 EUR
AATB100210O0 AATB100210O1
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 810)
Anticorpi
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Attenzione: For research use only.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: iFluor® 810
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR