Stampa…
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® 750)
Fornitore: AAT BIOQUEST
100810L0 100810L1
AATB100810L0EA 371 EUR
AATB100810L0 AATB100810L1
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® 750)
Anticorpi
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Attenzione: For research use only.
Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: iFluor® 750
Epitope:
Host: Mouse
Isotype:
Reactivity: Human