Stampa…
Bromocresol purple sodium salt 0,04% (w/v) in soluzione acquosa
Fornitore: Thermo Fisher Scientific
Synonyms: Sodium α-(3-bromo-5-methyl-4-oxo-2,5-cyclohexadienylidene)-α-(3-bromo-5-methyl-4-hydroxyphenyl)toluenesulphonate, Bromocresol purple water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-methylphenolate
38700.AE 38700.AP
38700.AEEA 31.3 EUR
38700.AE 38700.AP
Bromocresol purple sodium salt 0,04% (w/v) in soluzione acquosa
Bromocresol purple sodium salt
CAS No.: 62625-30-3 Molecular Formula: C21H15Br2NaO5S Formula Weight: 562.22 Physical Form: Liquid Appearance: Orange
Formula: C₂₁H₁₅Br₂NaO₅S
Peso molecolare: 562.21 g/mol
Punto di fusione: 255
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00148896
CAS Number: 62625-30-3
EINECS: 263-655-3