4-Metilstirene ≥98% stabilizzato
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
4-Viniltoluene
Stabilization:
Stabilised with 3,5-di-tert-butylpyrocatechol 0,10 %
Stabilized with 0.1% 3,5-di-tert-butylcatechol
Purity: 98+%
CAS No.: 622-97-9
Molecular Formula: C9H10
Formula Weight: 118.18
Storage Temperature: 2°C to 8°C
Flash Point: 45°(113°F)
Refractive Index: 1.5430
Melting Point: -34°
Boiling Point: 171-175°
Beilstein Reference: 1209317
UN No.: 2618
MDL No.: MFCD00008621
Formula:
C₉H₁₀ Peso molecolare: 118,18 g/mol Punto di ebollizione: 171…175 °C Punto di fusione: –34 °C Densità: 0,896 Flash Pt: 45 °C (113 °F) |
MDL Number:
MFCD00008621 CAS Number: 622-97-9 EINECS: 210-762-8 UN: 2618 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...