Stampa…

1,3-Diaminocyclohexane cis- and trans-mixture ≥95.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

C0813-25ML C0813.06 C0813-5ML
TCIAC0813-25MLEA 181 EUR
TCIAC0813-25ML FLUOC0813.06 TCIAC0813-5ML
1,3-Diaminocyclohexane cis- and trans-mixture ≥95.0% (by GC, titration analysis)
1,3-Diaminocyclohexane
Formula: C₆H₁₄N₂
Peso molecolare: 114,19 g/mol
Punto di ebollizione: 194°C
MDL Number: MFCD00059563
CAS Number: 3385-21-5
EINECS: 222-194-8
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a1948f4361028b7dd559e0a4eee9fe928454818&vid=1dee7c8e264e05c7c299e72a303b1077b085aa96&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 % (total of isomer)
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR