Stampa…

Anti-Rabbit IgG Goat Polyclonal Antibody (CF®583R)

Fornitore: Biotium
20913-50UL 20913-250UL
20913-50ULEA 103 EUR
20913-50UL 20913-250UL
Anti-Rabbit IgG Goat Polyclonal Antibody (CF®583R)
Anticorpi
Goat anti-rabbit IgG (H+L), F(ab’)2 fragment secondary antibody labeled with our superior CF® Dyes and other labels.

  • Available in 14 bright and photostable CF® Dyes, and HRP
  • Biotin, APC, and R-PE conjugates also available
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues

This is a goat anti-rabbit IgG (H L) F(ab’)2 fragment secondary antibody labeled with our superior CF® dyes and other labels.

Type: Secondary
Antigen: Rabbit IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Goat
Isotype: IgG (H+L)
Reactivity: Rabbit
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR