Stampa…
5-Nitro-6,7-dichloro-1,4-dihydro-2,3-quinoxalinedione ≥98% (by TLC)
Fornitore: ENZO LIFE SCIENCES AG
ALX550228M005 ALX550228M025
ENZOALX550228M005EA 170.5 EUR
ENZOALX550228M005 ENZOALX550228M025
5-Nitro-6,7-dichloro-1,4-dihydro-2,3-quinoxalinedione ≥98% (by TLC)
5-Nitro-6,7-dichloro-1,4-dihydro-2,3-quinoxalinedione
Potent and selective NMDA receptor glycine site antagonist.
Soluble in DMSO or dilute aqueous base.
Formula: C₈H₃Cl₂N₃O₄
Peso molecolare: 276,0 g/mol
Temperatura di stoccaggio: Refrigerata
CAS Number: 153504-81-5
Visualizza i codici
Purity (TLC, elemental analysis) ≥98%