Stampa…
(2R,4R)-APDC ≥98%
Fornitore: ENZO LIFE SCIENCES
ALX550178M001 ALX550178M005
ENZOALX550178M001EA 132 EUR
ENZOALX550178M001 ENZOALX550178M005
(2R,4R)-APDC ≥98%
(2R,4R)-APDC
mGluR agonist
Selective agonist for group II mGluRs. Inhibits the NADPH-oxidase dependent ROS system.
CAS Number: 169209-63-6