Stampa…
Gemcitabine hydrochloride ≥98%
Fornitore: ENZO LIFE SCIENCES
Synonyms: 2'-Deoxy-2',2'-difluorocytidine, dFdC, 4-Amino-1-((2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride
ALX480101M025 ALX480101M100
ENZOALX480101M025EA 149 EUR
ENZOALX480101M025 ENZOALX480101M100
Gemcitabine hydrochloride ≥98%
Gemcitabine hydrochloride
DNA synthesis inhibitor
Deoxycytidine analog with cytotoxic effects. Induces apoptosis and inhibits DNA synthesis.
Synthetic.
Formula: C₉H₁₂ClF₂N₃O₄
Peso molecolare: 299.6591 g/mol
MDL Number: MFCD01735988
CAS Number: 122111-03-9