Stampa…
Arabinofuranosyl-2-fluoroadenine ≥98% (by HPLC)
Fornitore: ENZO LIFE SCIENCES
Synonyms: 2-Fluoroadenine Arabinosid, (2R,3S,4S,5R)-2-(6-Amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 9-beta-D-Arabinofuranosyl-2-fluoro-9H-purin-6-amine, 9-beta-D-Arabinofuranosyl-2-fluoroadenine, Fludarabine
ALX480100M005 ALX480100M025
ENZOALX480100M005EA 83.6 EUR
ENZOALX480100M005 ENZOALX480100M025
Arabinofuranosyl-2-fluoroadenine ≥98% (by HPLC)
Arabinofuranosyl-2-fluoroadenine
F-ara-A precursor
Prodrug converted to the free nucleoside 9-beta-D-arabinosyl-2-fluoroadenine (F-ara-A). F-ara-A enters cells and accumulates as the 5'-triphosphate. Interferes with DNA synthesis and repair. Induces apoptosis of cancer cells. Inhibits RNA transcription.
Formula: C₁₀H₁₂FN₅O₄
Peso molecolare: 285.24 g/mol
MDL Number: MFCD00132942
CAS Number: 21679-14-1