Stampa…
Cicloesimide ≥95% (by TLC)
Fornitore: ENZO LIFE SCIENCES
Synonyms: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
ALX380269G001 ALX380269G005
ENZOALX380269G001EA 84.7 EUR
ENZOALX380269G001 ENZOALX380269G005
Cicloesimide ≥95% (by TLC)
Cicloesimide
Protein synthesis inhibitor
Antibiotic. Inhibits protein synthesis in eukaryotes by interfering with the translocation step. Induces apoptosis in tumor cell lines but can also delay or inhibit apoptosis induced by other agents. Widely used for selection of resistant strains of yeast and fungy, controlled inhibiton of protein synthesis for detecton of short-lived proteins and super-induction of protein expression. At 10µg/ml cycloheximide totally inhibits protein synthesis in mouse thymocytes.
Isolated from Streptomyces griseus.
Formula: C₁₅H₂₃NO₄
Peso molecolare: 281.35 g/mol
MDL Number: MFCD00082346
CAS Number: 66-81-9