Stampa…
Okadaic acid sodium salt
Fornitore: ENZO LIFE SCIENCES
Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin sodium salt, OA sodium salt
ALX350011C025 ALX350011C100 ALX350011M001
ENZOALX350011C025EA 112 EUR
ENZOALX350011C025 ENZOALX350011C100 ENZOALX350011M001
Okadaic acid sodium salt
Okadaic acid sodium salt
Formula: C₄₄H₆₇NaO₁₃
Peso molecolare: 827.00 g/mol
MDL Number: MFCD00210208
CAS Number: 209266-80-8