Stampa…
Compound E ≥98% (by HPLC)
Fornitore: ENZO LIFE SCIENCES AG
Synonyms: (S)-2-(2-(3,5-Difluorophenyl)acetamido)-N-((S)-1-methyl-2-oxo-5-phenyl-2,3-dihydro-1H-benzo[e][1,4]diazepin-3-yl)propanamide
ALX270415C250 ALX270415M001
ENZOALX270415C250EA 198 EUR
ENZOALX270415C250 ENZOALX270415M001
Compound E ≥98% (by HPLC)
Compound E
gamma-Secretase inhibitor
Cell permeable, potent, selective, non-transition state and non-competitive inhibitor of γ-secretase (IC50=0.3nM for total β-amyloid) and Notch processing. At higher concentrations (20-400µM), only weakly affects the presenilase activity. Activity tested!
Formula: C₂₇H₂₄F₂N₄O₃
Peso molecolare: 490,511 g/mol
MDL Number: MFCD04974531
CAS Number: 209986-17-4