Stampa…
Bisindolylmaleimide III ≥98%
Fornitore: ENZO LIFE SCIENCES
Synonyms: 3-(1-(3-Aminopropyl)-1H-indol-3-yl)-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione
ALX270051M001 ALX270051M005
ENZOALX270051M001EA 75.9 EUR
ENZOALX270051M001 ENZOALX270051M005
Bisindolylmaleimide III ≥98%
Bisindolylmaleimide III
PKC inhibitor
Potent and selective inhibitor of protein kinase C (PKC).
Formula: C₂₃H₂₀N₄O₂
Peso molecolare: 384,4 g/mol
MDL Number: MFCD01076633
CAS Number: 137592-43-9