Stampa…
Bisindolylmaleimide II ≥97% (by 1H-NMR)
Fornitore: ENZO LIFE SCIENCES
ALX270050M001 ALX270050M005
ENZOALX270050M001EA 83.6 EUR
ENZOALX270050M001 ENZOALX270050M005
Bisindolylmaleimide II ≥97% (by 1H-NMR)
Bisindolylmaleimide II
PKC inhibitor
Formula: C₂₇H₂₆N₄O₂
Peso molecolare: 438,5 g/mol
CAS Number: 137592-45-1