Stampa…
F(ab’)2 Anti-IgG Goat Polyclonal Antibody (CF®505)
Fornitore: Biotium
20916-250UL 20916-50UL
20916-250ULEA 165 EUR
20916-250UL 20916-50UL
F(ab’)2 Anti-IgG Goat Polyclonal Antibody (CF®505)
Anticorpi
This is a goat anti-mouse IgG (H L), F(ab’)2 fragment secondary antibody labeled with our superior CF® Dyes and other labels.
  • Available in 14 bright and photostable CF® Dyes, and HRP
  • Biotin, APC, and R-PE conjugates also available
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®505
Epitope:
Host: Goat
Isotype: IgG (H L)
Reactivity: Mouse