Stampa…
VU0155069 ≥98% (by TLC)
Fornitore: ENZO LIFE SCIENCES
Synonyms: (S)-N-(1-(4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-1-yl)piperidin-1-yl)propan-2-yl)-2-naphthamide
BMLST3700005 BMLST3700025
ENZOBMLST3700005EA 110 EUR
ENZOBMLST3700005 ENZOBMLST3700025
VU0155069 ≥98% (by TLC)
VU0155069
PLD inhibitor
Potent and specific inhibitor of phospholipase D1 (PLD1) in cellular (PLD1 IC50 = 11nM Calu-1 cells; PLD2 IC50 = 1800nM HEK293-gfpPLD2 cells) and in vitro (hPLD1 IC50 = 46nM; hPLD2 IC50 = 933nM) assays. Significantly decreases invasive migration in breast cancer cell lines (MDA-231, 4T1 and PMT).
Formula: C₂₆H₂₇ClN₄O₂
Peso molecolare: 462.970 g/mol
MDL Number: MFCD23160355
CAS Number: 1130067-06-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2afb982827a66b6096470732f092b6c3bad14547&vid=9afb32822a42afe34e785efdb07f7c7e9c80ec97&prid=web&clid=SAPDAM