Stampa…
KU-0063794 ≥99% (by HPLC)
Fornitore: ENZO LIFE SCIENCES AG
Synonyms: (5-(2-(cis-2,6-Dimethylmorpholino)-4-morpholinopyrido[2,3-d]pyrimidin-7-yl)-2-methoxyphenyl)methanol
ENZCHM1350005 ENZCHM1350010
ENZOENZCHM1350005EA 243.1 EUR
ENZOENZCHM1350005 ENZOENZCHM1350010
KU-0063794 ≥99% (by HPLC)
KU-0063794
Highly selective and potent inhibitor against both mTORC1 and mTORC2.
Formula: C₂₅H₃₁N₅O₄
Peso molecolare: 465,6 g/mol
MDL Number: MFCD22666594
CAS Number: 938440-64-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbaab94ae033e136eadef91ba92e2ada5403fceb&vid=e597c3acaa10f607f47855133b6df09f72cd520a&prid=web&clid=SAPDAM