Stampa…
Dasatinib ≥99% (by HPLC)
Fornitore: ENZO LIFE SCIENCES AG
Synonyms: N-(2-Chloro-6-methylphenyl)-2-((6-(4-(2-hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxamide
ENZCHM1520010 ENZCHM1520050
ENZOENZCHM1520010EA 72.6 EUR
ENZOENZCHM1520010 ENZOENZCHM1520050
Dasatinib ≥99% (by HPLC)
Dasatinib
Potent tyrosine kinase inhibitor
Dasatinib is an oral inhibitor of Bcr-Abl tyrosine-kinase, Src family tyrosine kinases, c-Kit, and ephrin receptors. It is being evaluated for use in numerous cancers, including advanced prostate cancer.
Formula: C₂₂H₂₆ClN₇O₂S
Peso molecolare: 488 g/mol
MDL Number: MFCD11046566
CAS Number: 302962-49-8