Stampa…

(±)-α-Acido Metossifenilacetico Reag. Ph. Eur. 1053601

Fornitore: VWR Chemicals

Synonyms: DL-α-Methoxyphenylacetic acid, 2-Methoxy-2-phenylacetic acid, α-Methoxyphenylacetic acid

87869.030EA 621 EUR
87869.030 87869.030DP
(±)-α-Acido Metossifenilacetico Reag. Ph. Eur. 1053601
(±)-α-Acido Metossifenilacetico
Formula: C₉H₁₀O₃
Peso molecolare: 166,18 g/mol
MDL Number: MFCD00064217
CAS Number: 7021-09-2
EINECS: 000-000-0
ADR: 3,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR