Methyl tridecanoate ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
Tridecylic acid methyl ester, Tridecanoic acid methyl ester
Formula:
C₁₄H₂₈O₂ Peso molecolare: 228.38 g/mol Punto di ebollizione: 163°C Punto di fusione: 7°C |
MDL Number:
MFCD00008977 CAS Number: 1731-88-0 EINECS: 217-054-8 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...