Stampa…

Tetratilammonio cloruro ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: Tetrylammonium chloride

T0095-25G T0095-500G T0095-100G
TCIAT0095-25GEA 25 EUR
TCIAT0095-25G TCIAT0095-500G TCIAT0095-100G
Tetratilammonio cloruro ≥98.0% (by titrimetric analysis)
Tetratilammonio cloruro
Formula: C₈H₂₀ClN
Peso molecolare: 165.71 g/mol
Punto di fusione: 285
MDL Number: MFCD00011828
CAS Number: 56-34-8
EINECS: 200-267-5
Merck Index: 14,09200
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85496c6fa73de3169f5a48f4850fa04a9b941a6a&vid=367ed227aba45b086f6cae884da6e4929272a12d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
pH 4.0 to 7.0 (50g/L, 25deg-C)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR