Stampa…

Phthalimide potassium salt ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-Potassium phthalimide, Potassium 1,3-dioxoisoindolin-2-ide, Potassium phthalimide

P0403-25G P0403-500G
TCIAP0403-25GEA 16 EUR
TCIAP0403-25G TCIAP0403-500G
Phthalimide potassium salt ≥98.0% (by titrimetric analysis)
Phthalimide potassium salt
Formula: C₈H₄KNO₂
Peso molecolare: 185,22 g/mol
Punto di fusione: > 300 °C
Densità: 1,634 g/cm³ (20 °C)
MDL Number: MFCD00005887
CAS Number: 1074-82-4
EINECS: 214-046-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=374222e739ae1c177e07378f0eb76072bec96029&vid=374222e739ae1c177e07378f0eb76072bec96029&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR