N-(4-Bromobutyl)phthalimide
Fornitore: Merck
Synonyms:
2-(4-Bromobutyl)isoindoline-1,3-dione
8.18222.0270
8.18222.0280
8.18222.0500
8.18222.0270EA
0
EUR
8.18222.0270
8.18222.0280
8.18222.0500
N-(4-Bromobutyl)phthalimide
N-(4-Bromobutyl)phthalimide
Formula:
C₁₂H₁₂BrNO₂ Peso molecolare: 282,14 g/mol Punto di fusione: 77…80 °C |
MDL Number:
MFCD00005905 CAS Number: 5394-18-3 EINECS: 226-401-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...