6-Chloro-3-methyluracil ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
C2300-25G
C2300-5G
TCIAC2300-25GEA
124
EUR
TCIAC2300-25G
TCIAC2300-5G
6-Chloro-3-methyluracil ≥98.0% (by HPLC, titration analysis)
6-Chloro-3-methyluracil
Formula:
C₅H₅ClN₂O₂ Punto di fusione: 278°C |
MDL Number:
MFCD01074837 CAS Number: 4318-56-3 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...