Stampa…

2-Deoxy-L-ribose

Fornitore: DEXTRA LABORATORIES

Synonyms: 2-Deoxy-L-erythropentose, 2-Deoxy-L-arabinose, 2-Deoxy-L(+)-ribose

BR52-1 BR52-2
DEXTBR52-1EA 0 EUR
DEXTBR52-1 DEXTBR52-2
2-Deoxy-L-ribose
2-Deoxy-L-ribose
Formula: C₅H₁₀O₄
Peso molecolare: 134,13 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00132941
CAS Number: 18546-37-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR