4-Butylphenyl isocyanate ≥98%
Fornitore: Thermo Scientific
|
Pericolo
|
Purity: 98%
CAS No.: 69342-47-8
Molecular Formula: C11H13NO
Formula Weight: 175.23
Storage Temperature: 2°C to 8°C
Flash Point: 108°(226°F)
Refractive Index: 1.5165
Boiling Point: 107-108°/3mm
Beilstein Reference: 390859
UN No.: 2206
MDL No.: MFCD00013882
Formula:
C₁₁H₁₃NO Peso molecolare: 175,23 g/mol Punto di ebollizione: 107…108 °C (3 mmHg) Densità: 0,992 Flash Pt: 108 °C (226 °F) |
MDL Number:
MFCD00013882 CAS Number: 69342-47-8 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...