Methyl isovalerate ≥98%
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
Isopropyl acetic acid methyl ester, Isovaleric acid methyl ester, 3-Methylbutyric acid emthyl ester, Methyl isopentanoate, 3-Methylbutanoic acid methyl ester
Purity: 98%
CAS No.: 556-24-1
Molecular Formula: (CH3)2CHCH2CO2CH3
Formula Weight: 116.17
Flash Point: 16°(60°F)
Refractive Index: 1.3936
Boiling Point: 115-117°
Beilstein Reference: 1699922
Merck Reference: 14,6093
UN No.: 2400
MDL No.: MFCD00042866
Formula:
C₆H₁₂O₂ Peso molecolare: 116,16 g/mol Punto di ebollizione: 115…117 °C Densità: 0,880 Flash Pt: 16 °C (60 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00042866 CAS Number: 556-24-1 EINECS: 209-117-3 UN: 2400 ADR: 3,II Merck Index: 14,6093 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...