N-Methylethylamine ≥98%
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
N-Ethylmethylamine
Purity: 98+%
CAS No.: 624-78-2
Molecular Formula: CH3CH2NHCH3
Formula Weight: 59.11
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₃H₉N Peso molecolare: 59,11 g/mol Punto di ebollizione: 33…34 °C Densità: 0,688 Flash Pt: <–34 °C (–29 °F) |
MDL Number:
MFCD00009030 CAS Number: 624-78-2 EINECS: 210-862-1 ADR: 3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...