2-Amino-3-chloropyridine 95%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 95%
CAS No.: 39620-04-7
Molecular Formula: C5H5ClN2
Formula Weight: 128.56
Melting Point: 59-62°
MDL No.: MFCD03541052
Formula:
C₅H₅ClN₂ Punto di fusione: 59…62 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03541052 CAS Number: 39620-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...