4-Iodopyridine 96%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 96%
CAS No.: 15854-87-2
Molecular Formula: C5H4IN
Formula Weight: 205.00
Storage Temperature: 2°C to 8°C
Melting Point: 96-99°
Beilstein Reference: 1594
MDL No.: MFCD02093937
Formula:
C₅H₄IN Peso molecolare: 205 g/mol Punto di fusione: 96…99 °C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD02093937 CAS Number: 15854-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...