Stampa…

5-Bromo-2-thenoic acid ≥98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 5-Bromothiophene-2-carboxylic acid, 5-Bromo-2-thiophenecarboxylic acid

A19242.03 A19242.06 A19242.14
A19242.03EA 30.1 EUR
A19242.03 A19242.06 A19242.14
5-Bromo-2-thenoic acid ≥98%
5-Bromo-2-thenoic acid

Purity: 98% CAS No.: 7311-63-9 Molecular Formula: C5H3BrO2S Formula Weight: 207.05 Melting Point: 139-143° Beilstein Reference: 118364

Formula: C₅H₃BrO₂S
Peso molecolare: 207,05 g/mol
Punto di fusione: 139…143 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00079725
CAS Number: 7311-63-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR