Stampa…

Isophytol 95%

Fornitore: Thermo Scientific

Synonyms: 3,7,11,15-Tetramethylhexadec-1-en-3-ol, 3,7,11,15-Tetramethylhexadec-1-en-3-ol

A18583.09 A18583.18
A18583.09EA 29.2 EUR
A18583.09 A18583.18
Isophytol 95%
Isophytol

Purity: 95% CAS No.: 505-32-8 Molecular Formula: C20H40O Formula Weight: 296.54 Refractive Index: 1.4571 Boiling Point: 125-128°/0.06mm Merck Reference: 14,5198 MDL No.: MFCD00048380

Formula: C₂₀H₄₀O
Peso molecolare: 296,54 g/mol
Punto di ebollizione: 125…128 °C (0,06 mmHg)
Densità: 0,8519
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00048380
CAS Number: 505-32-8
EINECS: 208-008-8
Merck Index: 14,5198

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR