Stampa…

2-Naphthalenesulphonyl chloride ≥97%

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: Naphthalene-2-sulphonyl chloride, β-Naphthalenesulphonyl chloride

A16872.14 A16872.22 A16872.36
A16872.14EA 52.7 EUR
A16872.14 A16872.22 A16872.36
2-Naphthalenesulphonyl chloride ≥97%
2-Naphthalenesulphonyl chloride

Purity: 97% CAS No.: 93-11-8 Molecular Formula: C10H7ClO2S Formula Weight: 226.68 Melting Point: 74-78° Boiling Point: 200-202°/13mm UN No.: 3261 MDL No.: MFCD00004087

Formula: C₁₀H₇ClO₂S
Punto di ebollizione: 200…202 °C (13 mmHg)
Punto di fusione: 74…78 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004087
CAS Number: 93-11-8
EINECS: 202-219-9
UN: 3261
ADR: 8,II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR