Isobutyl chloroformate ≥98%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Purity: 98%
CAS No.: 543-27-1
Molecular Formula: C5H9ClO2
Formula Weight: 136.58
Flash Point: 30°(86°F)
Refractive Index: 1.4060
Melting Point: -80°
Boiling Point: 128-129°
Beilstein Reference: 956590
Merck Reference: 14,5139
UN No.: 2742
MDL No.: MFCD00000642
Formula:
C₅H₉ClO₂ Punto di ebollizione: 128…129 °C Punto di fusione: –80 °C Densità: 1,048 Flash Pt: 30 °C (86 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00000642 CAS Number: 543-27-1 EINECS: 208-840-1 UN: 2742 ADR: 6.1,II Merck Index: 14,5139 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...