2-Chlorobenzoylacetonitrile 95%
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
3-(2-Chlorophenyl)-3-oxopropanenitrile
A13611.06
A13611.14
A13611.22
A13611.06EA
60.4
EUR
A13611.06
A13611.14
A13611.22
2-Chlorobenzoylacetonitrile 95%
2-Chlorobenzoylacetonitrile
Purity: 98%
CAS No.: 40018-25-5
Molecular Formula: C9H6ClNO
Formula Weight: 179.61
Melting Point: 54-58°
Beilstein Reference: 2413204
Formula:
C₉H₆ClNO Peso molecolare: 179,61 g/mol Punto di fusione: 54…58 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00051624 CAS Number: 40018-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...