Stampa…

4-Chlorobenzoylacetonitrile ≥98%

Fornitore: Thermo Scientific
Pericolo

Synonyms: 4-Chlorophenacylcyanide, 3-(4-Chlorophenyl)-3-oxopropanenitrile

A12364.06 A12364.14 A12364.22
A12364.06EA 65.3 EUR
A12364.06 A12364.14 A12364.22
4-Chlorobenzoylacetonitrile ≥98%
4-Chlorobenzoylacetonitrile

Purity: 98% CAS No.: 4640-66-8 Molecular Formula: C9H6ClNO Formula Weight: 179.61 Melting Point: 128-132° Beilstein Reference: 743368

Formula: C₉H₆ClNO
Peso molecolare: 179,61 g/mol
Punto di fusione: 128…132 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00051625
CAS Number: 4640-66-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR