4-Nitrophthalic acid 97%
Fornitore: Thermo Scientific
|
Attenzione
|
A10368.06
A10368.14
A10368.18
A10368.06EA
23.6
EUR
A10368.06
A10368.14
A10368.18
4-Nitrophthalic acid 97%
4-Nitrophthalic acid
Purity: 97%
CAS No.: 610-27-5
Molecular Formula: C8H5NO6
Formula Weight: 211.13
Odour: Odourless
Melting Point: 160-165°
Beilstein Reference: 1882262
MDL No.: MFCD00007252
Formula:
C₈H₅NO₆ Peso molecolare: 211,13 g/mol Punto di fusione: 160…165 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00007252 CAS Number: 610-27-5 EINECS: 210-215-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...