Methoxytrimethylsilane ≥97%
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
Trimethyl(methoxy)silane
Purity: 97+%
CAS No.: 1825-61-2
Molecular Formula: C4H12OSi
Formula Weight: 104.22
Flash Point: -11°(-12°F)
Physical Form: Liquid
Density: 0.756
Refractive Index: 1.3678
Sensitivity: Moisture Sensitive
Odour: Mild
Boiling Point: 57-58°
UN No.: 1993
MDL No.: MFCD00008344
Formula:
C₄H₁₂OSi Peso molecolare: 104,22 g/mol Punto di ebollizione: 57…58 °C Densità: 0,756 Flash Pt: –11 °C (–12 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008344 CAS Number: 1825-61-2 EINECS: 217-369-0 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...