Disodium tetrabromopalladate ≥99.95% (metals basis) Pd ≥ 22.0%, Premion®
Fornitore: Thermo Scientific
|
Attenzione
|
40407.03
40407.06
40407.09
40407.03EA
91.8
EUR
40407.03
40407.06
40407.09
Disodium tetrabromopalladate ≥99.95% (metals basis) Pd ≥ 22.0%, Premion®
Disodium tetrabromopalladate
Purity: 99.95%
CAS No.: 50495-13-1
Molecular Formula: Na2PdBr4
Formula Weight: 472.02
Physical Form: Powder
Formula:
Br₄Na₂Pd Peso molecolare: 472,02 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00046199 CAS Number: 50495-13-1 EINECS: 256-605-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...