Cesium hydrogen carbonate ≥99.99% (metals basis)
Fornitore: Thermo Scientific
14234.14
14234.22
14234.14EA
87.1
EUR
14234.14
14234.22
Cesium hydrogen carbonate ≥99.99% (metals basis)
Cesium hydrogen carbonate
Purity: 99.99%
CAS No.: 29703-01-3
Molecular Formula: CsHCO3
Formula Weight: 193.92
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
MDL No.: MFCD00010961
Formula:
CHCsO₃ Punto di fusione: 175 °C (dec.) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00010961 CAS Number: 15519-28-5 EINECS: 249-784-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...