Stampa…

Potassium tetranitroplatinate(II) (≥42,6% Pt)

Fornitore: Thermo Scientific

10538.03 10538.06
10538.03EA 134 EUR
10538.03 10538.06
Potassium tetranitroplatinate(II) (≥42,6% Pt)
Potassium tetranitroplatinate(II)

CAS No.: 13815-39-9 Molecular Formula: K2Pt(NO2)4 Formula Weight: 457.32 Physical Form: Crystalline MDL No.: MFCD00011375

Formula: K₂N₄O₈Pt
Peso molecolare: 457,3 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011375
CAS Number: 13815-39-9
EINECS: 237-491-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR